Learn More
MS436, MedChemExpress
Product Code. 16496366
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16496366
Brand: Medchem Express HY13959
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress MS436 is a new class of bromodomain inhibitor, exhibits potent affinity of an estimated Ki=30-50 nM for the BRD4 BrD1 and a 10-fold selectivity over the BrD2.
MS436 is a new class of bromodomain inhibitor, exhibits potent affinity of an estimated Ki=30-50 nM for the BRD4 BrD1 and a 10-fold selectivity over the BrD2.
Specifications
Chemical Name or Material MS436
CAS 1395084-25-9
Color Reddish Brown
Purity Grade Notes Research
Molecular Formula C18H17N5O3S
Quantity 100 mg
Solubility Information DMSO : 25.5 mg/mL (66.51 mM; Need ultrasonic and warming)
SMILES O=S(C1=CC=C(/N=N/C2=CC(C)=C(O)C=C2N)C=C1)(NC3=NC=CC=C3)=O
Molecular Weight (g/mol) 383.42
Formula Weight 383.42
Percent Purity 98.07%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death