missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse ASCL1/Mash1 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
225.00 € - 524.00 €
Specifications
| Antigen | ASCL1/Mash1 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, Immunohistochemistry 3-15 ug/mL |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18771218
![]() |
R&D Systems
AF2567-SP |
25 μg |
225.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18082692
![]() |
R&D Systems
AF2567 |
100 μg |
524.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ASCL1/Mash1 Polyclonal specifically detects ASCL1/Mash1 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| ASCL1/Mash1 | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 3-15 ug/mL | |
| Polyclonal | |
| Goat | |
| Mouse | |
| achaete-scute complex (Drosophila) homolog-like 1, achaete-scute complex homolog 1 (Drosophila), achaete-scute complex-like 1 (Drosophila), achaete-scute homolog 1, ASCL1, ASH1, ASH-1, ASH1MASH1, BHLHA46, bHLHa46achaete-scute complex-like 1, Class A basic helix-loop-helix protein 46, HASH1, HASH1achaete scute protein, Mash1 | |
| ASCL1 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| Q53WV9 | |
| 429 | |
| E. coli-derived recombinant mouse ASCL1/Mash1 His74-Asp173 (Gln160Glu) Accession # Q53WV9 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects mouse ASCL1/Mash1 in direct ELISAs and Western blots. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title