Learn More
Mouse ASCL1/Mash1 Antibody, R&D Systems™
GREENER_CHOICE
Goat Polyclonal Antibody has been used in 2 publications
Brand:  R&D Systems AF2567
 View more versions of this product
Product Code. 18082692
  • 524.00 € / 100µg
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
ASCL1/Mash1 Polyclonal specifically detects ASCL1/Mash1 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ASCL1/Mash1
Polyclonal
Unconjugated
Q53WV9
ASCL1
E. coli-derived recombinant mouse ASCL1/Mash1 His74-Asp173 (Gln160Glu) Accession # Q53WV9
100 μg
Primary
Detects mouse ASCL1/Mash1 in direct ELISAs and Western blots.
Mouse
IgG
Western Blot, Immunohistochemistry
LYOPH
Western Blot 0.1 ug/mL, Immunohistochemistry 3-15 ug/mL
achaete-scute complex (Drosophila) homolog-like 1, achaete-scute complex homolog 1 (Drosophila), achaete-scute complex-like 1 (Drosophila), achaete-scute homolog 1, ASCL1, ASH1, ASH-1, ASH1MASH1, BHLHA46, bHLHa46achaete-scute complex-like 1, Class A basic helix-loop-helix protein 46, HASH1, HASH1achaete scute protein, Mash1
Goat
Affinity Purified
RUO
429
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.