missing translation for 'onlineSavingsMsg'
Learn More

Monoethanolamine, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Product Code. p-5391234 Shop All J.T.Baker Products
Click to view available options
Quantity:
500 mL
Packaging:
Narrow Mouth Amber Glass Bottle
Unit Size:
500mL
This item is not returnable. View return policy

Product Code. 15548574

Brand: Avantor J.T.Baker 931401

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 141-43-5
Molecular Formula C2H7NO
Molecular Weight (g/mol) 61.08
MDL Number MFCD00008183
InChI Key HZAXFHJVJLSVMW-UHFFFAOYSA-N
Synonym ethanolamine, monoethanolamine, aminoethanol, 2-hydroxyethylamine, colamine, glycinol, olamine, 2-amino-1-ethanol, ethanol, 2-amino, ethylolamine
PubChem CID 700
ChEBI CHEBI:16000
IUPAC Name 2-aminoethan-1-ol
SMILES NCCO

Specifications

Flash Point 86.00°C
Packaging Narrow Mouth Amber Glass Bottle
Quantity 500 mL
Specific Gravity 1L = 1.02kg
Formula Weight 61.08
Grade BAKER ANALYZED™ ACS Reagent
Chemical Name or Material Monoethanolamine
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.