Learn More
Monensin sodium 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Monensin Sodium Salt |
| CAS | 22373-78-0 |
| Analyte or Component Names | Monensin Sodium Salt |
| Shipping Condition | Room Temperature |
| Molecular Formula | C36 H61 O11 . Na |
| Concentration | 100 μg/mL |
| Certifications/Compliance | ISO 17034 |
| Synonym | Monensin, sodium salt (1:1), 1,6-Dioxaspiro[4.5]decane-7-butyric acid, 2-[5-ethyltetrahydro-5-[tetrahydro-3-methyl-5-[tetrahydro-6-hydroxy-6-(hydroxymethyl)-3,5-dimethyl-2H-pyran-2-yl]-2-furyl]-2-furyl]-9-hydroxy-ÃŽ2-methoxy-ÃŽ±,ÃŽ3,2,8-tetramethyl-, monosodium salt (8CI), Monensin, monosodium salt (9CI), CRC Rumensin, Coban, Coban 45, Monensin A sodium salt, Monensin sodium, Monensin sodium salt, NSC 343257, Rumensin, Sodium monensin |
| InChI Formula | InChI=1S/C36H62O11.Na/c1-10-34(31-20(3)16-26(43-31)28-19(2)15-21(4)36(41,18-37)46-28)12-11-27(44-34)33(8)13-14-35(47-33)17-25(38)22(5)30(45-35)23(6)29(42-9)24(7)32(39)40;/h19-31,37-38,41H,10-18H2,1-9H3,(H,39,40);/q;+1/p-1/t19-,20-,21+,22+,23-,24-,25-,26+,27+,28-,29+,30-,31+,33-,34-,35+,36-;/m0./s1 |
| SMILES | [Na+].CC[C@]1(CC[C@@H](O1)[C@]2(C)CC[C@]3(C[C@H](O)[C@@H](C)[C@H](O3)[C@@H](C)[C@@H](OC)[C@H](C)C(=O)[O-])O2)[C@@H]4O[C@H](C[C@@H]4C)[C@H]5O[C@@](O)(CO)[C@H](C)C[C@@H]5C |
| Show More |