Learn More
Molybdenum Standard for ICP/MS, SPEX CertiPrep™
Product Code. 15265946 Shop All SPEX Certiprep Products
Click to view available options
Quantity:
125 mL
Unit Size:
125mL
Product Code. 15265946
Brand: SPEX Certiprep CLMO91BY
Please to purchase this item. Need a web account? Register with us today!
Includes
NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
AnalyteConcentration
Mo1μg/mL
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Packaging Triple-leached LDPE bottle
CAS 7439-98-7, 7732-18-5
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Analyte or Component Names Molybdenum Standard
Physical Form Fluid
Molecular Formula Mo
MDL Number MFCD00003465
Technique ICP-MS
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solution Type Standard Solution
InChI Key ZOKXTWBITQBERF-UHFFFAOYSA-N
Type ICP/ICP-MS Standard
SMILES [Mo]
Element Mo
IUPAC Name molybdenum
Solvent or Matrix Water
Molecular Weight (g/mol) 95.95
PubChem CID 23932
Quantity 125 mL
ChEBI CHEBI:28685
Grade Claritas PPT
Concentration or Composition (by Analyte or Components) 1mg/L
Chemical Name or Material Molybdenum Standard, ICP/MS
Show More Show Less
Product intended for laboratory use only