Learn More
MMP-2 (72kDa Collagenase IV) Ab-4 Mouse Monoclonal Antibody, Epredia™
Epredia™ MMP-2 (72kDa Collagenase IV) Ab-4, Mouse Monoclonal Antibody provides accurate, reproducible results.
130.56 € - 675.00 €
Specifications
| Antigen | MMP-2 (72kDa Collagenase IV) Ab-4 |
|---|---|
| Clone | A-Gel VC2 |
| Classification | Monoclonal |
| Form | Concentrated |
| Host Species | Mouse |
| Product Code | Brand | Quantity | Conjugate | Concentration | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Conjugate | Concentration | Price | Quantity & Availability | |||
|
12622357
|
Epredia
MS-806-B0 |
100 μL | Biotin | 200μg/mL | |||||
|
12632357
|
Epredia
MS-806-B1 |
500 μL | Biotin | 200μg/mL | |||||
|
12612357
|
Epredia
MS-806-B |
1 mL | Biotin | 200μg/mL | |||||
|
12652357
|
Epredia
MS-806-P0 |
100 μL | Unconjugated | 200μg/mL | |||||
|
12642357
|
Epredia
MS-806-P |
1 mL | Unconjugated | 200μg/mL | |||||
|
12672357
|
Epredia
MS-806-P1ABX |
100 μL | Unconjugated | 1mg/mL | |||||
|
12682357
|
Epredia
MS-806-PABX |
200 μL | Unconjugated | 1mg/mL | |||||
Description
MMPs are proteolytic enzymes capable of degrading connective tissue components. They have a common mode of activation, a conserved amino acid sequence in the putative metal binding-active site region, and are inhibited by specific tissue inhibitors of metalloproteinases (TIMPs). MMPs and TIMPs play a significant role in regulating angiogenesis. MMP-2; also known as 72kDa collagenase IV or gelatinase A is synthesized as a 631 amino acid proenzyme which is activated by cleavage of the first 80 amino acids.Host Species: Mouse
Clone: A-Gel VC2
Isotype: IgG1
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Human APMA-activated native 72kDa MMP-2
Molecular Weight: 72kDa (proform) and ∽66kDa (active form)
Positive Control: Placenta; Bladder, breast and ovarian CAs
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (not qualified)
Note:
USA: RUO; Int'l: RUO
Specifications
| MMP-2 (72kDa Collagenase IV) Ab-4 | |
| Monoclonal | |
| Mouse | |
| Proteases And Inhibitors | |
| Human APMA-activated native 72kDa MMP-2 | |
| Primary |
| A-Gel VC2 | |
| Concentrated | |
| RUO | |
| Human | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.