missing translation for 'onlineSavingsMsg'
Learn More

MMP-2 (72kDa Collagenase IV) Ab-4 Mouse Monoclonal Antibody, Epredia™

Epredia™ MMP-2 (72kDa Collagenase IV) Ab-4, Mouse Monoclonal Antibody provides accurate, reproducible results.

Brand:  Epredia MS-806-P1ABX

 View more versions of this product

Product Code. 12672357

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

MMPs are proteolytic enzymes capable of degrading connective tissue components. They have a common mode of activation, a conserved amino acid sequence in the putative metal binding-active site region, and are inhibited by specific tissue inhibitors of metalloproteinases (TIMPs). MMPs and TIMPs play a significant role in regulating angiogenesis. MMP-2; also known as 72kDa collagenase IV or gelatinase A is synthesized as a 631 amino acid proenzyme which is activated by cleavage of the first 80 amino acids.

Host Species: Mouse

Clone: A-Gel VC2

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Human APMA-activated native 72kDa MMP-2

Molecular Weight: 72kDa (proform) and ∽66kDa (active form)

Positive Control: Placenta; Bladder, breast and ovarian CAs

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (not qualified)

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

MMP-2 (72kDa Collagenase IV) Ab-4
A-Gel VC2
Unconjugated
Human APMA-activated native 72kDa MMP-2
RUO
Primary
Concentrated
Monoclonal
1mg/mL
Mouse
100 μL
Proteases And Inhibitors
Human
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.