Learn More
MMAE-d8, MedChemExpress
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
Description
MMAE-d8 (Monomethyl auristatin E-d8) is a deuterated labeled MMAE, a potent mitotic inhibitor and a tubulin inhibitor.
Specifications
Specifications
| Chemical Name or Material | MMAE-d8 |
| CAS | 2070009-72-0 |
| Purity Grade Notes | Research |
| Physical Form | Solid |
| Molecular Formula | C39H59D8N5O7 |
| Technique | Cancer-programmed Cell Death |
| Color | White |
| Synonym | Monomethyl auristatin E-d8 Deuterated labeled MMAE |
| Solubility Information | DMSO : 100 mg/mL (137.74 mM; Need ultrasonic) |
| SMILES | CC[C@H](C)[C@@H]([C@@H](CC(N1[C@@H](CCC1)[C@@H]([C@@H](C)C(N[C@H](C)[C@@H](O)C2=CC=CC=C2)=O)OC)=O)OC)N(C([C@@](C(C([2H])([2H])[2H])(C([2H])([2H])[2H])[2H])([2H])NC([C@H](C(C)C)NC)=O)=O)C |
| Show More |