Learn More
ML355, MedChemExpress
Product Code. 16487827
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16487827
Brand: Medchem Express HY123415mg
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress ML355 is a potent and selective inhibitor of 12-Lipoxygenase (12-LOX) with an IC50 of 0.34 μM, shows excellent selectivity over related lipoxygenases and cyclooxygenases, and possesses favorable ADME properties.
ML355 is a potent and selective inhibitor of 12-Lipoxygenase (12-LOX) with an IC50 of 0.34 μM, shows excellent selectivity over related lipoxygenases and cyclooxygenases, and possesses favorable ADME properties.
Specifications
Chemical Name or Material ML355
CAS 1532593-30-8
Color Yellow
Purity Grade Notes Research
Molecular Formula C21H19N3O4S2
Quantity 5 mg
Solubility Information DMSO : ≥ 42 mg/mL (95.13 mM)
SMILES O=S(NC1=NC2=CC=CC=C2S1)(C3=CC=C(NCC4=CC=CC(OC)=C4O)C=C3)=O
Molecular Weight (g/mol) 441.52
Formula Weight 441.52
Percent Purity 95.05%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation