Learn More
Mirtazapine EP Impurity B, Mikromol™
Description
Mirtazapine EP Impurity B
Specifications
Specifications
| Chemical Name or Material | (2-[(2RS)-4-Methyl-2-phenyl-piperazin-1-yl]pyridin-3-yl)methanol |
| CAS | 61337-89-1 |
| Analyte or Component Names | (2-[(2RS)-4-Methyl-2-phenyl-piperazin-1-yl]pyridin-3-yl)methanol |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H21 N3 O |
| Certifications/Compliance | ISO 17025 |
| Synonym | [2-[(2RS)-4-Methyl-2-phenylpiperazin-1-yl]pyridin-3-yl]methanol, [2-[(2RS)-4-Methyl-2-phenyl-piperazin-1-yl]pyridin-3-yl]methanol, Mirtazapine Imp. B (EP), Mirtazapine Impurity B |
| InChI Formula | InChI=1S/C17H21N3O/c1-19-10-11-20(17-15(13-21)8-5-9-18-17)16(12-19)14-6-3-2-4-7-14/h2-9,16,21H,10-13H2,1H3 |
| SMILES | CN1CCN(C(C1)c2ccccc2)c3ncccc3CO |
| IUPAC Name | [2-(4-methyl-2-phenylpiperazin-1-yl)pyridin-3-yl]methanol |
| Show More |