Learn More
Cytiva Microarray Hybridization Solution v2.0
Product Code. 10762047 Shop All Cytiva Products
Click to view available options
Quantity:
1 Piece
Unit Size:
Each
Product Code. 10762047
Brand: Cytiva RPK0325
Please to purchase this item. Need a web account? Register with us today!
Uses a blend of proprietary components to provide quality automated and manual slide hybridizations. Cytiva Microarray Hybridization Solution v2.0 features hybridization rate enhancers that improve the detection of rare targets, plus blocking agents that minimize nonspecific background.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Piece