missing translation for 'onlineSavingsMsg'
Learn More
Learn More
METTL7A Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77336-0.025mg
This item is not returnable.
View return policy
Description
METTL7A Polyclonal specifically detects METTL7A in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| METTL7A | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| AAM-B, DKFZP586A0522, EC 2.1.1, EC 2.1.1.-, methyltransferase like 7A, methyltransferase-like protein 7A, Protein AAM-B | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 25840 | |
| Human, Mouse | |
| IgG |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 2 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 2 μg/mL | |
| NP_054752 | |
| METTL7A | |
| Antibody was raised against a 13 amino acid synthetic peptide near the center of human MettL7A. The immunogen is located within amino acids 130 - 180 of MettL7A. Amino Acid Squence: SVKNQERILREV | |
| 0.025 mg | |
| Primary | |
| At least two isoforms of MettL7A are known to exist. This antibody is predicted to not cross-react with MettL7B. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction