missing translation for 'onlineSavingsMsg'
Learn More
Learn More
METTL7A Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
168.00 €
Specifications
| Antigen | METTL7A |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 2 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 2 μg/mL |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
Description
METTL7A Polyclonal specifically detects METTL7A in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| METTL7A | |
| Western Blot 2 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 2 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse | |
| NP_054752 | |
| 25840 | |
| Antibody was raised against a 13 amino acid synthetic peptide near the center of human MettL7A. The immunogen is located within amino acids 130 - 180 of MettL7A. Amino Acid Squence: SVKNQERILREV | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 1 mg/mL | |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| AAM-B, DKFZP586A0522, EC 2.1.1, EC 2.1.1.-, methyltransferase like 7A, methyltransferase-like protein 7A, Protein AAM-B | |
| METTL7A | |
| IgG | |
| Affinity Purified | |
| At least two isoforms of MettL7A are known to exist. This antibody is predicted to not cross-react with MettL7B. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title