Learn More
Methyl trans-Cinnamate, Mikromol™
Product Code. 30050080
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30050080 100 mg 100mg
30049974 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30050080 Supplier Mikromol Supplier No. MM0370.01
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Methyl trans-Cinnamate
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Methyl trans-Cinnamate
CAS 1754-62-7
Analyte or Component Names Methyl trans-Cinnamate
Shipping Condition Room Temperature
Molecular Formula C10 H10 O2
Certifications/Compliance ISO 17025
Synonym Methyl trans-Cinnamate
InChI Formula InChI=1S/C10H10O2/c1-12-10(11)8-7-9-5-3-2-4-6-9/h2-8H,1H3/b8-7+
SMILES COC(=O)\C=C\c1ccccc1
IUPAC Name methyl (E)-3-phenylprop-2-enoate
Molecular Weight (g/mol) 162.19
Quantity 100 mg
Formula Weight 162.0681 g/mol