missing translation for 'onlineSavingsMsg'
Learn More

Methyl trans-3-nonenoate, 98%, Thermo Scientific Chemicals

Product Code. 11469546
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11469546 25 g 25g
11459546 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11469546 Supplier Thermo Scientific Alfa Aesar Supplier No. A19288.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36781-67-6
Molecular Formula C10H18O2
Molecular Weight (g/mol) 170.25
MDL Number MFCD00065419
InChI Key MTDCXFZGUVZRSQ-BQYQJAHWSA-N
Synonym methyl 3-nonenoate, 3-nonenoic acid, methyl ester, methyl trans-3-nonenoate, methyl e-non-3-enoate, unii-yu6w67i34w, methyl 3-nonenoate, 3e, fema no. 3710, methyl non-3-enoate, 3-nonenoic acid, methyl ester, e, 3-nonenoic acid, methyl ester, 3e
PubChem CID 5366844
IUPAC Name methyl (E)-non-3-enoate
SMILES CCCCC\C=C\CC(=O)OC

Specifications

Density 0.89
Boiling Point 216°C to 217°C
Flash Point 177°C (351°F)
Refractive Index 1.436
Quantity 25 g
Formula Weight 170.25
Percent Purity 98%
Chemical Name or Material Methyl trans-3-nonenoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.