accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COOCH3(E)

CAS RN 36781-67-6

IUPAC Name: methyl (3E)-non-3-enoate
Synonyms: methyl 3-nonenoate
Molecular Weight (g/mol): 170.25
Molecular Formula: C10H18O2
InChi Key: MTDCXFZGUVZRSQ-BQYQJAHWSA-N
SMILES: CCCCC\C=C\CC(=O)OC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results