Learn More
Methyl Toluene-4-sulphonate, Mikromol™
Product Code. 17920526
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17920526 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17920526 Supplier Mikromol Supplier No. MM0722.01
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Methyl Toluene-4-sulphonate
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Methyl Toluene-4-sulphonate
CAS 80-48-8
Analyte or Component Names Methyl Toluene-4-sulphonate
Shipping Condition Room Temperature
Molecular Formula C8 H10 O3 S
Certifications/Compliance ISO 9001
Synonym Methyl Toluene-4-sulphonate
InChI Formula InChI=1S/C8H10O3S/c1-7-3-5-8(6-4-7)12(9,10)11-2/h3-6H,1-2H3
SMILES COS(=O)(=O)c1ccc(C)cc1
IUPAC Name methyl 4-methylbenzenesulfonate
Molecular Weight (g/mol) 186.23
Quantity 100 mg
Formula Weight 186.0351 g/mol