missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methyl Red hydrochloride, ACS

Product Code. p-7050384
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11314187

Brand: Thermo Scientific Chemicals 036668.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 63451-28-5 | C15H16ClN3O2 | 305.762 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63451-28-5
Molecular Formula C15H16ClN3O2
Molecular Weight (g/mol) 305.762
MDL Number MFCD00012614
InChI Key ICTWKXAVJAHRMK-UHFFFAOYSA-N
Synonym 2-[4-(Dimethylamino)phenyl]azobenzoic acid
PubChem CID 113227
IUPAC Name 2-[[4-(dimethylamino)phenyl]diazenyl]benzoic acid;hydrochloride
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)O.Cl

Specifications

Melting Point 175°C (decomposition)
Formula Weight 305.77
Grade ACS
Quantity 25 g
Physical Form Crystalline
Chemical Name or Material Methyl Red hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.