accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 63451-28-5

HClH3CNCH3NNOHO(E/Z)

CAS RN 63451-28-5

IUPAC Name: hydrogen 2-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzoic acid chloride
Synonyms: hydrogen methyl red chloride
Molecular Weight (g/mol): 305.76
Molecular Formula: C15H16ClN3O2
InChi Key: ICTWKXAVJAHRMK-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CN(C)C1=CC=C(C=C1)N=NC1=CC=CC=C1C(O)=O
Grade 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Color 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)