Learn More
Methyl Quinoxaline-2-carboxylate, Mikromol™
Product Code. 30049267
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30049267 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30049267 Supplier Mikromol Supplier No. MM3695.010025
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Methyl Quinoxaline-2-carboxylate
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Methyl Quinoxaline-2-carboxylate
CAS 1865-11-8
Analyte or Component Names Methyl Quinoxaline-2-carboxylate
Shipping Condition Room Temperature
Molecular Formula C10 H8 N2 O2
Certifications/Compliance ISO 9001
Synonym 2-Quinoxalinecarboxylic Acid Methyl Ester, Methyl Quinoxaline-2-carboxylate
InChI Formula InChI=1S/C10H8N2O2/c1-14-10(13)9-6-11-7-4-2-3-5-8(7)12-9/h2-6H,1H3
SMILES COC(=O)c1cnc2ccccc2n1
IUPAC Name methyl quinoxaline-2-carboxylate
Molecular Weight (g/mol) 188.18
Quantity 25 mg
Formula Weight 188.0586 g/mol