Learn More
Methyl 5-(2-Bromoacetyl)-2-hydroxybenzoate, Mikromol™
Description
Methyl 5-(2-Bromoacetyl)-2-hydroxybenzoate
Specifications
Specifications
| Chemical Name or Material | Methyl 5-(2-Bromoacetyl)-2-hydroxybenzoate |
| CAS | 36256-45-8 |
| Analyte or Component Names | Methyl 5-(2-Bromoacetyl)-2-hydroxybenzoate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C10 H9 Br O4 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Benzoic acid, 5-(2-bromoacetyl)-2-hydroxy-, methyl ester, Benzoic acid, 5-(bromoacetyl)-2-hydroxy-, methyl ester (9CI), Salicylic acid, 5-(bromoacetyl)-, methyl ester (7CI), 5-(2-Bromoacetyl)-2-hydroxybenzoic acid methyl ester, 5-(Bromoacetyl)salicylic acid methyl ester, Methyl 5-(2-Bromoacetyl)-2-hydroxybenzoate, Methyl 5-(bromoacetyl)-2-hydroxybenzoate, Methyl 5-(bromoacetyl)salicylate, 3-Carbomethoxy-4-hydroxy-alpha-bromoacetophenone |
| InChI Formula | InChI=1S/C10H9BrO4/c1-15-10(14)7-4-6(9(13)5-11)2-3-8(7)12/h2-4,12H,5H2,1H3 |
| SMILES | COC(=O)c1cc(ccc1O)C(=O)CBr |
| IUPAC Name | methyl 5-(2-bromoacetyl)-2-hydroxybenzoate |
| Show More |