Learn More
Methyl 5-[2-[Benzyl(tert-butyl)amino]acetyl]-2-hydroxybenzoate Hydrochloride, Mikromol™
Description
Methyl 5-[2-[Benzyl(tert-butyl)amino]acetyl]-2-hydroxybenzoate Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Methyl 5-[2-[Benzyl(tert-butyl)amino]acetyl]-2-hydroxybenzoate Hydrochloride |
| CAS | 27475-26-9 |
| Analyte or Component Names | Methyl 5-[2-[Benzyl(tert-butyl)amino]acetyl]-2-hydroxybenzoate Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H25 N O4 . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | Benzoic acid, 5-[2-[(1,1-dimethylethyl)(phenylmethyl)amino]acetyl]-2-hydroxy-, methyl ester, hydrochloride (1:1), Benzoic acid, 5-[[(1,1-dimethylethyl)(phenylmethyl)amino]acetyl]-2-hydroxy-, methyl ester, hydrochloride (9CI), Salicylic acid, 5-(N-benzyl-N-tert-butylglycyl)-, methyl ester, hydrochloride (8CI), 5-(N-Benzyl-N-tert-butylglycyl)salicylic acid methyl ester hydrochloride, Methyl 5-[2-[Benzyl(tert-butyl)amino]acetyl]-2-hydroxybenzoate Hydrochloride |
| InChI Formula | InChI=1S/C21H25NO4.ClH/c1-21(2,3)22(13-15-8-6-5-7-9-15)14-19(24)16-10-11-18(23)17(12-16)20(25)26-4;/h5-12,23H,13-14H2,1-4H3;1H |
| SMILES | Cl.COC(=O)c1cc(ccc1O)C(=O)CN(Cc2ccccc2)C(C)(C)C |
| IUPAC Name | methyl 5-[2-[benzyl(tert-butyl)amino]acetyl]-2-hydroxybenzoate;hydrochloride |
| Show More |