Learn More
Methyl 4-(2-Hydroxyethoxy)benzoate, Mikromol™
Product Code. 30458774
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30458774 4 x 25 mg 100mg
30051206 100 mg 100mg
30050148 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30458774 Supplier Mikromol Supplier No. MM0738.0600254AMP
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Methyl 4-(2-Hydroxyethoxy)benzoate, Mikromol
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Methyl 4-(2-Hydroxyethoxy)benzoate
CAS 3204-73-7
Analyte or Component Names Methyl 4-(2-Hydroxyethoxy)benzoate
Shipping Condition Room Temperature
Molecular Formula C10 H12 O4
Certifications/Compliance ISO 9001
Synonym Benzoic acid, 4-(2-hydroxyethoxy)-, methyl ester, Benzoic acid, p-(2-hydroxyethoxy)-, methyl ester (6CI,7CI,8CI), 4-(2-Hydroxyethoxy)benzoic acid methyl ester, Methyl 4-(2-hydroxyethoxy)benzoate, Methyl 4-(2'-hydroxyethoxy)benzoate, Methyl 4-(β-hydroxyethoxy)benzoate, Methyl p-(2-hydroxyethoxy)benzoate, Methyl p-(β-hydroxyethoxy)benzoate
InChI Formula InChI=1S/C10H12O4/c1-13-10(12)8-2-4-9(5-3-8)14-7-6-11/h2-5,11H,6-7H2,1H3
Solution Type Neat
SMILES COC(=O)c1ccc(OCCO)cc1
IUPAC Name methyl 4-(2-hydroxyethoxy)benzoate
Molecular Weight (g/mol) 196.2
Quantity 4 x 25 mg
Formula Weight 196.0736 g/mol