Learn More
Methyl 4-(2-Hydroxyethoxy)benzoate, Mikromol™
Description
Methyl 4-(2-Hydroxyethoxy)benzoate
Specifications
Specifications
| Chemical Name or Material | Methyl 4-(2-Hydroxyethoxy)benzoate |
| CAS | 3204-73-7 |
| Analyte or Component Names | Methyl 4-(2-Hydroxyethoxy)benzoate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C10 H12 O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Benzoic acid, 4-(2-hydroxyethoxy)-, methyl ester, Benzoic acid, p-(2-hydroxyethoxy)-, methyl ester (6CI,7CI,8CI), 4-(2-Hydroxyethoxy)benzoic acid methyl ester, Methyl 4-(2-hydroxyethoxy)benzoate, Methyl 4-(2'-hydroxyethoxy)benzoate, Methyl 4-(β-hydroxyethoxy)benzoate, Methyl p-(2-hydroxyethoxy)benzoate, Methyl p-(β-hydroxyethoxy)benzoate |
| InChI Formula | InChI=1S/C10H12O4/c1-13-10(12)8-2-4-9(5-3-8)14-7-6-11/h2-5,11H,6-7H2,1H3 |
| SMILES | COC(=O)c1ccc(OCCO)cc1 |
| IUPAC Name | methyl 4-(2-hydroxyethoxy)benzoate |
| Show More |