Learn More
Methyl 3-(thien-2-yl)acrylate, 97%, Thermo Scientific™
Product Code. 11469606
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11469606 5 g 5g
11459606 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11469606 Supplier Thermo Scientific Alfa Aesar Supplier No. B20177.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 20883-96-9
Molecular Formula C8H8O2S
Molecular Weight (g/mol) 168.21
MDL Number MFCD00173772
InChI Key HKVOGMDMMCLQFJ-SNAWJCMRSA-N
Synonym methyl 3-2-thienyl acrylate, methyl 3-thien-2-yl acrylate, methyl 3-thien-2-yl-acrylate, methyl e-3-thiophen-2-ylprop-2-enoate, methyl 2e-3-thiophen-2-yl prop-2-enoate, methyl 3-thiophen-2-yl prop-2-enoate, thiophene-2-acrylic acid methyl ester, e-methyl 3-thiophen-2-yl acrylate, methyl 2e-3-2-thienyl-2-propenoate, methyl 2e-3-2-thienyl prop-2-enoate
PubChem CID 5709013
IUPAC Name methyl (E)-3-thiophen-2-ylprop-2-enoate
SMILES COC(=O)C=CC1=CC=CS1
Specifications
Melting Point 48°C to 49°C
Quantity 5 g
Formula Weight 168.22
Percent Purity 97%
Chemical Name or Material Methyl 3-(thien-2-yl)acrylate