accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 20883-96-9

CH3OOS(E)

CAS RN 20883-96-9

IUPAC Name: methyl (2E)-3-(thiophen-2-yl)prop-2-enoate
Synonyms: methyl (2E)-3-(thiophen-2-yl)prop-2-enoate
Molecular Weight (g/mol): 168.21
Molecular Formula: C8H8O2S
InChi Key: HKVOGMDMMCLQFJ-SNAWJCMRSA-N
SMILES: COC(=O)\C=C\C1=CC=CS1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results