Learn More
Methyl (2RS)-(2-Chlorophenyl)[4,7-dihydrothieno[2,3-c]pyridin-6(5H)-yl]acetate Hydrochloride, Mikromol™
Description
Methyl (2RS)-(2-Chlorophenyl)[4,7-dihydrothieno[2,3-c]pyridin-6(5H)-yl]acetate Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Methyl (2-Chlorophenyl)[4,7-dihydrothieno[2,3-c]pyridin-6(5H)-yl]acetate Hydrochloride |
| CAS | 144750-52-7 |
| Analyte or Component Names | Methyl (2-Chlorophenyl)[4,7-dihydrothieno[2,3-c]pyridin-6(5H)-yl]acetate Hydrochloride |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C16 H16 Cl N O2 S . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Thieno[2,3-c]pyridine-6(5H)-acetic acid, α-(2-chlorophenyl)-4,7-dihydro-, methyl ester, hydrochloride (1:1), Thieno[2,3-c]pyridine-6(5H)-acetic acid, α-(2-chlorophenyl)-4,7-dihydro-, methyl ester, hydrochloride (9CI), Thieno[2,3-c]pyridine-6(5H)-acetic acid, α-(2-chlorophenyl)-4,7-dihydro-, methyl ester, hydrochloride, (±)-, Methyl (2RS)-(2-Chlorophenyl)[4,7-dihydrothieno[2,3-c]pyridin-6(5H)-yl]acetate hydrochloride, Methyl(±)-(o-chlorophenyl)-4,5-dihydrothieno[2,3-c]pyridine-6(7H)-acetate hydrochloride, Clopidogrel Related Compound B, Clopidogrel Related Compound B as Hydrochloride, Clopidogrel Hydrogen Sulfate Impurity B as racemic Hydrochloride, Clopidogrel Impurity B as racemic Hydrochloride |
| InChI Formula | InChI=1S/C16H16ClNO2S.ClH/c1-20-16(19)15(12-4-2-3-5-13(12)17)18-8-6-11-7-9-21-14(11)10-18;/h2-5,7,9,15H,6,8,10H2,1H3;1H |
| SMILES | Cl.COC(=O)C(N1CCc2ccsc2C1)c3ccccc3Cl |
| IUPAC Name | methyl 2-(2-chlorophenyl)-2-(5,7-dihydro-4H-thieno[2,3-c]pyridin-6-yl)acetate;hydrochloride |
| Show More |