Learn More
Methyl 2-(4-Fluorophenyl)-2-(piperidin-2-yl)acetate Hydrochloride, TRC
Product Code. 30481882
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30481882 1 mg 1mg
30474131 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30481882 Supplier Toronto Research Chemicals Supplier No. TRCM2723901MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Methyl 2-(4-Fluorophenyl)-2-(piperidin-2-yl)acetate
CAS 2732991-86-3
Analyte or Component Names Methyl 2-(4-Fluorophenyl)-2-(piperidin-2-yl)acetate
Shipping Condition Room Temperature
Molecular Formula C14 H18 F N O2 . Cl H
Synonym α-(4-Fluorophenyl)-2-piperidineacetic Acid Methyl Ester Hydrochloride
InChI Formula InChI=1S/C14H18FNO2.ClH/c1-18-14(17)13(12-4-2-3-9-16-12)10-5-7-11(15)8-6-10;/h5-8,12-13,16H,2-4,9H2,1H3;1H
Solution Type Neat
SMILES Cl.COC(=O)C(C1CCCCN1)c2ccc(F)cc2
IUPAC Name methyl 2-(4-fluorophenyl)-2-(2-piperidyl)acetate;hydrochloride
Industry Type Forensics
Molecular Weight (g/mol) 287.76
Quantity 1 mg
Formula Weight 287.1088