Learn More
Methyl 2-[2,3-Dichloro-4-[2-(methoxymethyl)butanoyl]phenoxy]acetate, Mikromol™
Product Code. 30052034
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30052034 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30052034 Supplier Mikromol Supplier No. MM1207.070025
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Methyl 2-[2,3-Dichloro-4-[2-(methoxymethyl)butanoyl]phenoxy]acetate
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Methyl 2-[2,3-Dichloro-4-[2-(methoxymethyl)butanoyl]phenoxy]acetate
CAS 2726534-39-8
Analyte or Component Names Methyl 2-[2,3-Dichloro-4-[2-(methoxymethyl)butanoyl]phenoxy]acetate
Shipping Condition Room Temperature
Molecular Formula C15 H18 Cl2 O5
Certifications/Compliance ISO 9001
Synonym Methyl 2-[2,3-Dichloro-4-[2-(methoxymethyl)butanoyl]phenoxy]acetate
InChI Formula InChI=1S/C15H18Cl2O5/c1-4-9(7-20-2)15(19)10-5-6-11(14(17)13(10)16)22-8-12(18)21-3/h5-6,9H,4,7-8H2,1-3H3
SMILES CCC(COC)C(=O)c1ccc(OCC(=O)OC)c(Cl)c1Cl
IUPAC Name methyl 2-[2,3-dichloro-4-[2-(methoxymethyl)butanoyl]phenoxy]acetate
Molecular Weight (g/mol) 349.21
Quantity 25 mg
Formula Weight 348.0531 g/mol