missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl [1-(4-Chlorobenzoyl)-5-methoxy-2-methyl-1H-indol-3-yl]acetate (Indomethacin Methyl Ester), Mikromol™
Description
Methyl [1-(4-Chlorobenzoyl)-5-methoxy-2-methyl-1H-indol-3-yl]acetate (Indomethacin Methyl Ester)
Specifications
Specifications
| Chemical Name or Material | Indomethacin Methyl Ester |
| CAS | 1601-18-9 |
| Analyte or Component Names | Indomethacin Methyl Ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H18 Cl N O4 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 1-(4-Chlorobenzoyl)-5-methoxy-2-methyl-1H-indole-3-acetic acid methyl ester, Indole-3-acetic acid, 1-(p-chlorobenzoyl)-5-methoxy-2-methyl-, methyl ester (7CI,8CI), Indomethacin methyl ester, L 588983, Methyl 1-(4-chlorobenzoyl)-5-methoxy-2-methylindole-3-acetate, Methyl N-(p-chlorobenzoyl)-2-methyl-5-methoxy-3-indolylacetate, Indometacin Imp. H (EP), Indometacin Impurity H |
| InChI Formula | InChI=1S/C20H18ClNO4/c1-12-16(11-19(23)26-3)17-10-15(25-2)8-9-18(17)22(12)20(24)13-4-6-14(21)7-5-13/h4-10H,11H2,1-3H3 |
| SMILES | COC(=O)Cc1c(C)n(C(=O)c2ccc(Cl)cc2)c3ccc(OC)cc13 |
| IUPAC Name | methyl 2-[1-(4-chlorobenzoyl)-5-methoxy-2-methylindol-3-yl]acetate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?