Learn More
Methyl [1-(4-Chlorobenzoyl)-5-methoxy-2-methyl-1H-indol-3-yl]acetate (Indomethacin Methyl Ester), Mikromol™
Product Code. 30052080
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Product Code. 30052080
Brand: Mikromol MM0584.08
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Methyl [1-(4-Chlorobenzoyl)-5-methoxy-2-methyl-1H-indol-3-yl]acetate (Indomethacin Methyl Ester)
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Indomethacin Methyl Ester
CAS 1601-18-9
Analyte or Component Names Indomethacin Methyl Ester
Shipping Condition Room Temperature
Molecular Formula C20 H18 Cl N O4
Certifications/Compliance ISO 17025
Synonym 1-(4-Chlorobenzoyl)-5-methoxy-2-methyl-1H-indole-3-acetic acid methyl ester, Indole-3-acetic acid, 1-(p-chlorobenzoyl)-5-methoxy-2-methyl-, methyl ester (7CI,8CI), Indomethacin methyl ester, L 588983, Methyl 1-(4-chlorobenzoyl)-5-methoxy-2-methylindole-3-acetate, Methyl N-(p-chlorobenzoyl)-2-methyl-5-methoxy-3-indolylacetate, Indometacin Imp. H (EP), Indometacin Impurity H
InChI Formula InChI=1S/C20H18ClNO4/c1-12-16(11-19(23)26-3)17-10-15(25-2)8-9-18(17)22(12)20(24)13-4-6-14(21)7-5-13/h4-10H,11H2,1-3H3
SMILES COC(=O)Cc1c(C)n(C(=O)c2ccc(Cl)cc2)c3ccc(OC)cc13
IUPAC Name methyl 2-[1-(4-chlorobenzoyl)-5-methoxy-2-methylindol-3-yl]acetate
Molecular Weight (g/mol) 371.81
Quantity 100 mg
Formula Weight 371.0924 g/mol