Learn More
Methisazone, LoGiCal
Product Code. 30457389
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30457389 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30457389 Supplier LoGiCal Supplier No. LGCFOR1916.00
Please to purchase this item. Need a web account? Register with us today!
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Methisazone
CAS 1910-68-5
Analyte or Component Names Methisazone
Shipping Condition Room Temperature
Molecular Formula C10 H10 N4 O S
Certifications/Compliance ISO 9001
Synonym Hydrazinecarbothioamide, 2-(1,2-dihydro-1-methyl-2-oxo-3H-indol-3-ylidene)-, Indole-2,3-dione, 1-methyl-, 3-(thiosemicarbazone) (7CI,8CI), Isatin, 1-methyl-, 3-(thiosemicarbazone) (6CI), 1-Methylindole-2,3-dione 3-(thiosemicarbazone), 1-Methylisatin 3-thiosemicarbazone, 1-Methylisatin β-thiosemicarbazone, 33T57, BW 33T57, Compound 33T57, Isathizone, Kemoviran, MIBT, Marboran, Methisazone, Metisazon, Metisazone, Metisazonum, N-Methylisatin 3-thiosemicarbazone, N-Methylisatin thiosemicarbazone, N-Methylisatin-β-thiosemicarbazone, NSC 69811, NSC 70969, Sch 16, Viruzona, β-Thiosemicarbazone-N-methylisatin
InChI Formula InChI=1S/C10H10N4OS/c1-14-7-5-3-2-4-6(7)8(9(14)15)12-13-10(11)16/h2-5H,1H3,(H3,11,13,16)/b12-8-
Solution Type Neat
SMILES CN1C(=O)\C(=N/NC(=S)N)\c2ccccc12
IUPAC Name [(Z)-(1-methyl-2-oxoindol-3-ylidene)amino]thiourea
Industry Type Clinical Testing
Molecular Weight (g/mol) 234.28
Quantity 5 mg
Formula Weight 234.0575 g/mol