Learn More
Methicillin sodium salt, 90+%, Thermo Scientific™
Brand:  Thermo Scientific Alfa Aesar J66414.03
Quantity 1g
Product Code. 15477759
  • ONLINE EXCLUSIVE PRICE
    282.00 €
    269.00 € / 1g
    Save 13.00 €
    4% Off
Please to purchase this item. Need a web account? Register with us today!
Description
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Targets primarily the cell wall of Gram-positive organisms especially Staphylococcus aureus. Methicillin is used to study bacterium susceptibility, and to inhibit cell-wall synthesis. PBP′s are enzymes which catalyze a pentagl
TRUSTED_SUSTAINABILITY
Specifications
Specifications
132-92-3
1g
Light sensitive
MFCD07787409
14,5969
Slightly soluble in acetone; Soluble in ethanol or water
[Na+].COC1=CC=CC(OC)=C1C(=O)N[C@H]1[C@H]2SC(C)(C)[C@@H](N2C1=O)C([O-])=O
402.40
CHEBI:52065
98%
White
>110°C (230°F)
C17H19N2NaO6S
640°C
Sodium (2,6-dimethoxyphenyl)penicillin; Sodium methicillin
MGFZNWDWOKASQZ-UMLIZJHQSA-M
sodium (2S,5R,6R)-6-(2,6-dimethoxybenzamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
23667627
402.4
Methicillin sodium salt
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Methicillin sodium salt
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Meticillin sodium
SDS Documents