accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 132-92-3

NaCH3OOCH3ONH(R)(R)SCH3CH3(S)NOOO

CAS RN 132-92-3

IUPAC Name: sodium (2S,5R,6R)-6-(2,6-dimethoxybenzamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
Synonyms: methicillin sodium methicillin sodium anhydrous meticillin sodium meticillin sodium salt dimocillin sodium
Molecular Weight (g/mol): 402.4
Molecular Formula: C17H19N2NaO6S
InChi Key: MGFZNWDWOKASQZ-UMLIZJHQSA-M
SMILES: [Na+].COC1=CC=CC(OC)=C1C(=O)N[C@H]1[C@H]2SC(C)(C)[C@@H](N2C1=O)C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Color 
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)