Learn More
Methenamine N-Oxide, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
Methenamine N-Oxide
Specifications
Specifications
| Chemical Name or Material | Methenamine N-Oxide |
| CAS | 62190-92-5 |
| Analyte or Component Names | Methenamine N-Oxide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C6 H12 N4 O |
| Certifications/Compliance | ISO 9001 |
| Synonym | 1,3,5,7-Tetraazatricyclo[3.3.1.13,7]decane, 1-oxide, Methenamine N-Oxide |
| InChI Formula | InChI=1S/C6H12N4O/c11-10-4-7-1-8(5-10)3-9(2-7)6-10/h1-6H2 |
| SMILES | [O-][N+]12CN3CN(CN(C3)C1)C2 |
| Molecular Weight (g/mol) | 156.19 |
| Show More |