Learn More
Phenthoate in Methanol 100μg/mL, Fisher Chemical™
Product Code. 12966114 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12966114 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12966114 Supplier Fisher Chemical Supplier No. P877820
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Specifications
Packaging Ampoule
CAS 2597-03-7
Analyte or Component Names Phenthoate
Molecular Formula C12H17O4PS2
Concentration 100 μg/mL
InChI Key XAMUDJHXFNRLCY-UHFFFAOYNA-N
Type Single-component Standard
SMILES CCOC(=O)C(C1=CC=CC=C1)SP(=S)(OC)OC
Element Phenthoate
IUPAC Name ethyl 2-dimethoxyphosphinothioylsulfanyl-2-phenylacetate
Solvent or Matrix Methanol
Molecular Weight (g/mol) 320.36
PubChem CID 17435
Quantity 1 mL
ChEBI CHEBI:34917