Learn More
Chlorpyrifos D10 in n-Nonane 100μg/mL, Fisher Chemical™
Product Code. 12972524 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12972524 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12972524 Supplier Fisher Chemical Supplier No. P812710
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Packaging Ampoule(s)
CAS 285138-81-0
Molecular Formula C9H11Cl3NO3PS
Concentration 100 μg/mL
InChI Key SBPBAQFWLVIOKP-MWUKXHIBSA-N
SMILES CCOP(=S)(OCC)OC1=NC(=C(C=C1Cl)Cl)Cl
IUPAC Name bis(1,1,2,2,2-pentadeuterioethoxy)-sulfanylidene-(3,5,6-trichloropyridin-2-yl)oxy-$l^{5}-phosphane
Solvent or Matrix n-Nonane
Molecular Weight (g/mol) 360.636
PubChem CID 16213399
Quantity 1 mL
Chemical Name or Material Chlorpyrifos D10