Learn More
Bromacil in Acetonitrile 100μg/mL, Fisher Chemical™
BIOPHARMA
Product Code. 12939663 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12939663 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12939663 Supplier Fisher Chemical Supplier No. P807460
Please to purchase this item. Need a web account? Register with us today!
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Specifications
Packaging Ampoule(s)
CAS 314-40-9
Molecular Formula C9H13BrN2O2
Concentration 100 μg/mL
UN Number 1648
InChI Key CTSLUCNDVMMDHG-UHFFFAOYSA-N
SMILES CCC(C)N1C(=O)C(=C(NC1=O)C)Br
IUPAC Name 5-bromo-3-butan-2-yl-6-methyl-1H-pyrimidine-2,4-dione
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 261.119
PubChem CID 9411
Quantity 1 mL
ChEBI CHEBI:83658
Chemical Name or Material Bromacil
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Standard solution Bromacil 100ug/ml in acetonitrile
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H311-Toxic in contact with skin.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P330-Rinse mouth.
  • P361 + P364-Take off immediately all contaminated clothing and wash it before reuse.
  • P501-Dispose of contents/container to .