Learn More
Metabolite Benzo(a)anthracenei n Methyl-tert.butylether 100μg/mL, Fisher Chemical™
Product Code. 12999603 Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Product Code. 12999603
Brand: Fisher Chemical P806420
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Packaging Ampoule(s)
CAS 1718-53-2
Molecular Formula C18H12
Concentration 100 μg/mL
UN Number 1145
InChI Key DXBHBZVCASKNBY-AQZSQYOVSA-N
SMILES C1=CC=C2C(=C1)C=CC3=CC4=CC=CC=C4C=C32
IUPAC Name 1,2,3,4,5,6,7,8,9,10,11,12-dodecadeuteriobenzo[a]anthracene
Solvent or Matrix Methyl-tert.butylether
Molecular Weight (g/mol) 240.367
PubChem CID 12279531
Quantity 1 mL
Chemical Name or Material Benzo(a)anthracene D12