Learn More
Mesalazine Methyl Ester, Mikromol™
Product Code. 30051671
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30051671 25 mg 25mg
30051032 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30051671 Supplier Mikromol Supplier No. MM0217.250025
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Mesalazine Methyl Ester
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Mesalazine Methyl Ester
CAS 42753-75-3
Analyte or Component Names Mesalazine Methyl Ester
Shipping Condition Room Temperature
Molecular Formula C8 H9 N O3
Certifications/Compliance ISO 17025
Synonym 5-Amino-2-hydroxybenzoic acid methyl ester, Methyl 2-hydroxy-5-aminobenzoate, Methyl 5-amino-2-hydroxybenzoate, Methyl 5-aminosalicylate, Methyl m-aminosalicylate
InChI Formula InChI=1S/C8H9NO3/c1-12-8(11)6-4-5(9)2-3-7(6)10/h2-4,10H,9H2,1H3
SMILES COC(=O)c1cc(N)ccc1O
IUPAC Name methyl 5-amino-2-hydroxybenzoate
Molecular Weight (g/mol) 167.16
Quantity 25 mg
Formula Weight 167.0582 g/mol