Learn More
Mesalazine Methyl Ester, Mikromol™
Description
Mesalazine Methyl Ester
Specifications
Specifications
| Chemical Name or Material | Mesalazine Methyl Ester |
| CAS | 42753-75-3 |
| Analyte or Component Names | Mesalazine Methyl Ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C8 H9 N O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 5-Amino-2-hydroxybenzoic acid methyl ester, Methyl 2-hydroxy-5-aminobenzoate, Methyl 5-amino-2-hydroxybenzoate, Methyl 5-aminosalicylate, Methyl m-aminosalicylate |
| InChI Formula | InChI=1S/C8H9NO3/c1-12-8(11)6-4-5(9)2-3-7(6)10/h2-4,10H,9H2,1H3 |
| SMILES | COC(=O)c1cc(N)ccc1O |
| IUPAC Name | methyl 5-amino-2-hydroxybenzoate |
| Show More |