Learn More
Mephosfolan 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Product Code. 30324132
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30324132 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30324132 Supplier Dr. Ehrenstorfer Supplier No. DREA14870000AL100
Please to purchase this item. Need a web account? Register with us today!
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Mephosfolan
CAS 950-10-7
Analyte or Component Names Mephosfolan
Shipping Condition Room Temperature
Molecular Formula C8 H16 N O3 P S2
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
InChI Formula InChI=1S/C8H16NO3PS2/c1-4-11-13(10,12-5-2)9-8-14-6-7(3)15-8/h7H,4-6H2,1-3H3/b9-8-
SMILES CCOP(=O)(OCC)\N=C/1\SCC(C)S1
IUPAC Name (Z)-N-diethoxyphosphoryl-4-methyl-1,3-dithiolan-2-imine
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 269.32
Quantity 1 mL
Formula Weight 269.0309 g/mol