missing translation for 'onlineSavingsMsg'
Learn More

Meloxicam, MedChemExpress

Product Code. 16442076
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16442076

Brand: Medchem Express HYB0261

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Meloxicam is a non-steroidal antiinflammatory agent, inhibits COX activity, with IC50s of 0.49 μM and 36.6 μM for COX-2 and COX-1, respectively.

Meloxicam is a non-steroidal antiinflammatory agent, inhibits COX activity, with IC50s of 0.49 μM and 36.6 μM for COX-2 and COX-1, respectively.

Specifications

Chemical Name or Material Meloxicam
CAS 71125-38-7
Color Yellowish Green
Purity Grade Notes Research
Molecular Formula C14H13N3O4S2
Quantity 500 mg
Solubility Information DMSO : 50 mg/mL (142.29 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(C1=C(O)C2=CC=CC=C2S(N1C)(=O)=O)NC3=NC=C(C)S3
Molecular Weight (g/mol) 351.4
Formula Weight 351.4
Percent Purity 99.88%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.