Learn More
Megestrol Acetate, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Megestrol Acetate
Specifications
Specifications
| Chemical Name or Material | Megestrol-17-acetate |
| CAS | 595-33-5 |
| Analyte or Component Names | Megestrol-17-acetate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C24 H32 O4 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Medroxyprogesterone Acetate Imp. G (EP), Megestrol Acetate, 6-Methyl-3,20-dioxopregna-4,6-dien-17-yl Acetate, 17-(Acetyloxy)-6-methylpregna-4,6-diene-3,20-dione, 17-Hydroxy-6-methylpregna-4,6-diene-3,20-dione acetate, 17-Acetoxy-6-methylpregna-4,6-diene-3,20-dione, 17alpha-Acetoxy-6-dehydro-6-methylprogesterone, 17alpha-Acetoxy-6-methylpregna-4,6-diene-3,20-dione, 17alpha-Hydroxy-6-methylpregna-4,6-diene-3,20-dione acetate, 5071, 6-Dehydro-6-methyl-17alpha-acetoxyprogesterone, 6-Methyl-17alpha-acetoxy-4,6-pregnadiene-3,20-dione, 6-Methyl-17alpha-hydroxy-Delta6-progesterone acetate, 6-Methyl-6-dehydro-17alpha-acetoxyprogesterone, 6-Methyl-Delta4,6-pregnadien-17alpha-ol-3,20-dione acetate, BDH 1298, DMAP, MGA, Magestin, Maygace, Megace, Megalia, Megeron, Megestat, Megestil, Megestin, Megestrol acetate, Megestryl acetate, NSC 71423, Nia, Niagestin, Ovaban, Ovarid, SC 10363, Medroxyprogesterone Acetate Impurity G |
| InChI Formula | InChI=1S/C24H32O4/c1-14-12-18-19(22(4)9-6-17(27)13-21(14)22)7-10-23(5)20(18)8-11-24(23,15(2)25)28-16(3)26/h12-13,18-20H,6-11H2,1-5H3/t18-,19+,20+,22-,23+,24+/m1/s1 |
| SMILES | CC(=O)O[C@@]1(CC[C@H]2[C@@H]3C=C(C)C4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C)C(=O)C |
| IUPAC Name | [(8R,9S,10R,13S,14S,17R)-17-acetyl-6,10,13-trimethyl-3-oxo-2,8,9,11,12,14,15,16-octahydro-1H-cyclopenta[a]phenanthren-17-yl] acetate |
| Show More |