Learn More
Mefenamic Acid Isopropyl Ester, Mikromol™
Product Code. 17933976
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17933976 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17933976 Supplier Mikromol Supplier No. MM0327.090025
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Mefenamic Acid Isopropyl Ester
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Mefenamic Acid Isopropyl Ester
CAS 2732673-37-7
Analyte or Component Names Mefenamic Acid Isopropyl Ester
Shipping Condition Room Temperature
Molecular Formula C18 H21 N O2
Certifications/Compliance ISO 9001
Synonym Mefenamic Acid Isopropyl Ester
InChI Formula InChI=1S/C18H21NO2/c1-12(2)21-18(20)15-9-5-6-10-17(15)19-16-11-7-8-13(3)14(16)4/h5-12,19H,1-4H3
SMILES CC(C)OC(=O)c1ccccc1Nc2cccc(C)c2C
IUPAC Name propan-2-yl 2-(2,3-dimethylanilino)benzoate
Molecular Weight (g/mol) 283.36
Quantity 25 mg
Formula Weight 283.1572 g/mol