Learn More
Mebeverine Hydrochloride, Mikromol™
Description
Mebeverine Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Mebeverine Hydrochloride |
| CAS | 2753-45-9 |
| Analyte or Component Names | Mebeverine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C25 H35 N O5 . Cl H |
| Certifications/Compliance | ISO 17034 |
| Synonym | Mebeverine Hydrochloride, 3,4-Dimethoxybenzoic acid 4-[ethyl[2-(4-methoxyphenyl)-1-methylethyl]amino]butyl ester hydrochloride, 4'-[N-Ethyl-1'-methyl-2'-(4''-methoxyphenyl)ethylamino]butyl 3,4-dimethoxybenzoate hydrochloride, CSAG 144, Colofac, Colospasmin, Duspatal, Duspatalin, Mebeverine hydrochloride, NSC 169101, 4-[Ethyl(p-methoxy-alpha-methylphenethyl)amino]veratric acid butyl ester hydrochloride, 4-[Ethyl(p-methoxy-alpha-methylphenethyl)amino]-1-butanol veratrate hydrochloride, 4-[Ethyl(p-methoxy-alpha-methylphenethyl)amino]butyl veratrate hydrochloride |
| InChI Formula | InChI=1S/C25H35NO5.ClH/c1-6-26(19(2)17-20-9-12-22(28-3)13-10-20)15-7-8-16-31-25(27)21-11-14-23(29-4)24(18-21)30-5;/h9-14,18-19H,6-8,15-17H2,1-5H3;1H |
| SMILES | Cl.CCN(CCCCOC(=O)c1ccc(OC)c(OC)c1)C(C)Cc2ccc(OC)cc2 |
| IUPAC Name | 4-[ethyl-[1-(4-methoxyphenyl)propan-2-yl]amino]butyl 3,4-dimethoxybenzoate;hydrochloride |
| Show More |