missing translation for 'onlineSavingsMsg'
Learn More

MBP Antibody, Novus Biologicals™

Goat Polyclonal Antibody

328.00 € - 526.00 €

Specifications

Antigen MBP
Concentration 1 mg/mL
Dilution Western Blot 1:5000-10000, Immunocytochemistry/ Immunofluorescence 1:2000-5000
Applications Western Blot, Immunofluorescence
Classification Polyclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18616006
View Documents
Novus Biologicals
NBP3-05553-100ul
100 μg
526.00 €
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18609487
View Documents
Novus Biologicals
NBP3-05553-50ul
50 μg
328.00 €
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

MBP Polyclonal antibody specifically detects MBP in Human, Mouse, Rat, Porcine, Bovine samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications

Specifications

MBP
Western Blot 1:5000-10000, Immunocytochemistry/ Immunofluorescence 1:2000-5000
Polyclonal
Purified
RUO
Human, Mouse, Rat, Pig, Bovine
MGC99675, Myelin A1 protein, myelin basic protein, Myelin membrane encephalitogenic protein
Purified MBP isolated from bovine brain
Immunogen affinity purified
1 mg/mL
Western Blot, Immunofluorescence
Unconjugated
Goat
Hypoxia Signaling, Immune System Diseases, Immunology, Neuroscience, Neurotransmission, Oligodendrocyte Cell Markers, Stem Cell Markers
50% PBS, 50% glycerol
4155
Primary
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.