missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MBP Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP3-05553-50ul
This item is not returnable.
View return policy
Description
MBP Polyclonal antibody specifically detects MBP in Human, Mouse, Rat, Porcine, Bovine samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| MBP | |
| Polyclonal | |
| Unconjugated | |
| 50% PBS, 50% glycerol | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Pig, Bovine | |
| Purified |
| Western Blot, Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 1:5000-10000, Immunocytochemistry/ Immunofluorescence 1:2000-5000 | |
| MGC99675, Myelin A1 protein, myelin basic protein, Myelin membrane encephalitogenic protein | |
| Purified MBP isolated from bovine brain | |
| 50 μg | |
| Hypoxia Signaling, Immune System Diseases, Immunology, Neuroscience, Neurotransmission, Oligodendrocyte Cell Markers, Stem Cell Markers | |
| 4155 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction