missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MATH2/NEUROD6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
234.00 € - 748.00 €
Specifications
| Antigen | MATH2/NEUROD6 |
|---|---|
| Dilution | Western Blot 1:200-1:1000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18194112
|
Novus Biologicals
NBP2-29865-0.025MG |
0.025 mg |
234.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18142494
|
Novus Biologicals
NBP2-29865 |
0.1 mg |
748.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MATH2/NEUROD6 Polyclonal specifically detects MATH2/NEUROD6 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| MATH2/NEUROD6 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| 50% saturated ammonium sulfate and PBS with No Preservative | |
| Atoh2, BHLHA2, bHLHa2MATH2, Class A basic helix-loop-helix protein 2, Math-2, NeuroD6, neurogenic differentiation 6, neurogenic differentiation factor 6, NEX1M, Protein atonal homolog 2 | |
| NEUROD6 | |
| IgG | |
| Affinity Purified | |
| Recognizes NeuroD 6 (MATH2, NEX1), a Helix-loop-helix class of transcription factor. |
| Western Blot 1:200-1:1000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse | |
| Q96NK8 | |
| 63974 | |
| Synthetic peptide. | |
| Primary | |
| Store at -70°C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title